SC-3 System Frocks, WORKLON®
Fournisseur: Superior Uniform
Certificats
A propos de cet article
Reusable frocks are designed for use in microelectronics, semiconductor, disk drive, laser, and other ISO Class 6 (FED-STD-209E Class 1000/M4.5) industrial cleanrooms.
- Available with snap front closure or zipper front closure
- Color-coded hang loop denotes size
The Burlington C3™ high density fabric dissipates static charges and guards against airborne particles and lint. Reusable cleanroom frocks have a two-ply reinforced placket, antistatic knit cuffs, and adjustable snaps at the collar.
Spécifications
- Catalog No:
- 14228-014
- 14228-016
- 14228-010
- 14228-012
- 14227-978
- 14228-022
- 14228-018
- 14228-020
- 14227-976
- 14228-006
- 14228-008
- 14228-004
- 14227-982
- 14227-992
- 14227-994
- 14227-988
- 14227-990
- 14228-000
- 14228-002
- 14227-996
- 14227-998
- 14227-984
- 14228-028
- 14227-986
- 14228-024
- 14228-026
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Risk area:
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Zip closure
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Zip closure
- Zip closure
- Zip closure
- Press studs / snaps
- Press studs / snaps
- Zip closure
- Press studs / snaps
- Press studs / snaps
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps