LD-100 System Frocks, 3454 Series, WORKLON®
Fournisseur: Superior Uniform
Certificats
A propos de cet article
Reusable cleanroom frocks are designed for use in pharmaceutical, medical device, food processing, and other industries where particulate control and bacteria filtration are required.
Frocks are constructed of 100% polyester taffeta with polyester knit cuffs, raglan sleeves, and adjustable snaps at the collar
Suitable for ISO Class 6 (FED-STD-209E Class 1000/M4.5) controlled environments.
A color-coded hang loop denotes size. Frocks have a duo closure style that features a zipper front closure as well as one lower snap. Two-ply reinforced placket. Gamma compatible. Autoclavable.
Spécifications
- Catalog No:
- 14228-058
- 14228-060
- 14228-054
- 14228-056
- 14228-044
- 14228-050
- 14228-052
- 14228-046
- 14228-048
- Model:
- 3454
- 3454
- 3454
- 3454
- 3454
- 3454
- 3454
- 3454
- 3454
- Garment application:
- Medical, Food, Clinical, Cleanroom, Laboratory
- Medical, Food, Clinical, Cleanroom, Laboratory
- Medical, Food, Clinical, Cleanroom, Laboratory
- Medical, Food, Clinical, Cleanroom, Laboratory
- Medical, Food, Clinical, Cleanroom, Laboratory
- Medical, Food, Clinical, Cleanroom, Laboratory
- Medical, Food, Clinical, Cleanroom, Laboratory
- Medical, Food, Clinical, Cleanroom, Laboratory
- Medical, Food, Clinical, Cleanroom, Laboratory
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester