A propos de cet article
Use to wipe, clean, or dry cuvettes, delicate glassware, syringes, pipets, and pH electrodes.
Use to wipe, clean, or dry cuvettes, delicate glassware, syringes, pipets, and pH electrodes. They are ideal for filtering and cleaning.
- Available in convenient dispenser box or on economical bolt
- Inert to even harsh solvents
- Mesh size: grade 10
Cheesecloth wipers are ideal for filtering and cleaning
Cheesecloth wipers are ideal for filtering and cleaning
Wipers meet cleanliness specifications of USP and Federal Specification CCC-G-101c.
Bleached, all-purpose cheesecloth wipers are brilliant white and certified 100% pure cotton of reagent-grade quality and purity. They feature a unique combination of cleanliness, softness, absorbency, and strength. Will not scratch the most sensitive surfaces. Absorb six times their weight and absorb both solvents and aqueous solutions.