Synthware Vacuum/Gas Adapter, Ball Joint, Kemtech America
# de catalogue 60000-170
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Description:Synthware Vacuum/Gas Adapter
- Cat. no.:60000-170
- Supplier No.:A293520
Spécifications
A propos de cet article
Vacuum/gas adapter with a ball joint.
- [ST] ball joint
- With hose connection
Featuring a [ST] 35/20 ball joint and 8mm hose connection.
Customers Who Bought This Also Bought