Clean & Safe® Hand Wipes, CleanTex™
# de catalogue 58019-522
Fournisseur: Advantus
Spécifications
- W×L:14 x 20.3 cm (51/2 x 8")
- Cat. no.:58019-522
- Supplier No.:CT818
Spécifications
A propos de cet article
Reduces bacteria on hands after contact with contaminated surfaces and materials.
Premoistened cleansing pads combine a highly effective antimicrobial agent (benzethonium chloride) in a cleansing and refreshing hydro-alcoholic solution.
Customers Who Bought This Also Bought