OpticPad® Optical Surface Cleaning Pads, CleanTex™
# de catalogue 58019-512
Fournisseur: Advantus
Spécifications
- W×L:14 x 20.3 cm (51/2 x 8")
- Cat. no.:58019-512
- Packaging:100 Pads per Box
- Supplier No.:CT811
Spécifications
A propos de cet article
Disposable, lint-free pad is presaturated for cleaning optical-grade surfaces.
Ideal for cleaning and polishing front and back surfaces of mirrors, microscope lenses, safety goggles, and photographic equipment. Solvent is filtered to 0.2 µm and evaporates quickly without leaving a residue.
Packaging: Each pad is hermetically sealed and individually wrapped.
Customers Who Bought This Also Bought