Spécifications
- Sterility:Sterile
- W×L:45.7×66 cm (18×26")
- Color:Blue
- Description:Nonabsorbent drape towel, dexter
- Cat. no.:75880-962
- Packaging:1 Towel/pack, 120 packs/case
- Supplier No.:7552
Spécifications
A propos de cet article
All-purpose towels are ideal for general laboratory cleanup or protection of cleanroom surfaces and equipment.
- Soft non woven tissue hand towels are reinforced with scrim laminate
Drape towels are made of either polylined material, which creates an impervious bottom, or non woven non absorbent wet-laid dexter material with an adhesive strip.