VWR® Spec-Wipe® 4 Sterile Wiper
Catalog # 21914-772
Supplier: VWR International
Small Business Enterprise
Specifications
- Basis weight:136 g/m²
- Absorbency:312 ml/m²
- Absorbency rate:≤1.5 sec.
- Extractables:0.042 g/m² using deionized water, 0.015 g/m² using isopropyl alcohol
- Fibres (>100 µm):3.3×10³/m²
- Particle generation (>=0,5 µm):4.8 x 10⁶/m²
- Ions - chloride:0.45 ppm
- Ions - sodium:0.39 ppm
- Cat. no.:21914-772
- W×L:30.5 x 30.5 cm (12 x 12")
Specifications
About this item
Wiper offers superior quality for aseptic environments where bacteria, particulate, and extractable levels are critical concerns.
- 100% double-knit polyester
- Continuous filament
- Superior quality for aseptic environments
This 100% double-knit polyester wiper has low levels of contaminants as a result of its tightly spun, continuous-filament material and clean-cut edges.
While designed to be durable for rigorous cleaning, it is soft enough for delicate surfaces. Wiper is laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom and sterilized by gamma radiation to a Sterility Assurance Level of 10-⁶ in accordance with ANSI/ISO/AAMI guidelines.
Certifications: Laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Packaging: 20 wipers per bag. 15 bags per case.
Customers Who Bought This Also Bought