SilSat™ Silicone-Impregnated Wiper, Micronova
# de catalogue 68400-782
Fournisseur: Micronova
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- W×L:10.2 x 15.2 cm (4 x 6")
- Material:NovaPoly
- Supplier No.:SSW6-01IR
Spécifications
A propos de cet article
Presaturated wiper provides lubricant in controlled applications.
- Gamma irradiated
Each wiper features isopropyl alcohol blended with several grams of medical grade silicone for applying to service gears and rollers on fill lines and other maintenance applications. Suitable for medical device, food processing, maintenance, and other applications requiring sterile lubrication.
Ordering information: Packed eight wipers per pouch.
Customers Who Bought This Also Bought