PROSAT® Presaturated Nonwoven Wipers, Contec®
Fournisseur: Contec
Certificats
A propos de cet article
Nonwoven wipers designed for cleaning laminar flow workbenches, cleanroom surfaces, and equipment.
Ideal for wiping articles prior to pass through. Compatible with ISO Class 5 (FED-STD-209E Class 100/M3.5) environments. Choose spunlaced 100% polyester wipers for improved cleanliness, meltblown polypropylene wipers for improved durability, or economical hydroentangled polyester/cellulose wipers.
Ordering information: Packaged in resealable pouches to preserve cleanliness and solvent saturation levels. Pouches are individually lot-numbered for traceability.
Spécifications
- Catalog No:
- 89428-544
- CA89074-010
- Basis weight:
- 36 g/m²
- 34 g/m²
- Extractables - using deionised water:
- 0.01 g/m²
- 0.040 g/m²
- Extractables - using isopropyl alcohol:
- 0.07 g/m²
- 0.020 g/m²
- Fibres (>100 µm):
- 6.3×10³/m²
- 0.145×10³/m²
- Particle generation (>=0,5 µm):
- 5.1 × 10⁶/m²
- 23.9 x 10⁶/m²
- Ions - chloride:
- 5 ppm
- 8.6 ppm
- Ions - sodium:
- 3.0 ppm
- 0.08 ppm