About this item
Containers for the disposal of radioactive waste can also be used with disposable NRC-approved waste bags
Constructed of durable 13mm (1/2") cast acrylic for maximum protection. Available with hinged or flip-top lids. Container IBWH-280-M has a handle. Containers with flip-top lids have a 2cm hole for disposal of contaminated pipets.