KIMAX® Cylindrical Weighing Bottles, Regular and Tall Form, Kimble®, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
With airtight, cap-style glass [ST] stopper.
- Area for marking on each bottle
- Capacity is to base of neck
Designed from Federal Specification NNN-1493, Type I, Style II requirements.
Customers Who Bought This Also Bought