Oily Waste Cans, Polyethylene, Eagle Manufacturing
Supplier: Eagle Manufacturing
Certificates
About this item
Cans are designed for the disposal of solvent and oily waste, rags, and other materials subject to spontaneous combustion.
- Impact-resistant
- Corrosion-resistant
- Rustproof
Impervious to many liquid chemicals, solvents, and corrosives. A foot lever raises the lid which closes automatically after use.
FM approved.
Customers Who Bought This Also Bought