Hybridization Bottles, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
These bottles are ideal for nucleic acid and protein blotting procedures that use a minimum of probe solution.
These borosilicate bottles are produced with a heavy wall for added safety and are available with a slip-resistant safety coating. The 35 mm bottles incorporate a fluoropolymer resin/silicone lined cap to prevent leakage. Both bottles incorporate a polypropylene cap with a welded fluoropolymer resin/silicone liner. Optional acrylic storage rack holds up to six hybridization bottles.
They are a safe and efficient alternative to messy bags, and allow hybridization and wash solutions to be quickly poured in and out. Bottles fit most popular hybridization ovens to simplify procedures such as Southern, Northern, Western, and slot blots.
Customers Who Bought This Also Bought