A propos de cet article
The Touch Free™ Automatic Waste Can’s motion-sensitive lid opens automatically to accept waste, and then closes into a fitted well that helps to reduce odor and airborne contamination.
Available in three slim, modern designs. Item 74950-064, White 2.5 gallon can is perfect for bench-top use and includes a removable rigid liner that won’t leak or rust. Body and lid are molded of easy-to-clean ABS plastic, and the removable pail is molded of autoclavable polypropylene. Use with 14 x 9" bags. Item 74950-066, Gray and 89125-226, Red 7.3 gallon cans have a molded polypropylene pail and gray ABS plastic top. Use with 24 x 30", 24 x 36" or 25 x 35" bags. Item 74950-068, 12 gallon can has a Stainless Steel body and black ABS plastic lid. Use with 24 x 30", 24 x 36" or 25 x 35" bags.
- Durable and dependable
- On-off switch deactivates the auto-open/close function
- Up to 4000 openings per set of batteries (not included)
The Touch Free™ Automatic Waste Can’s motion-sensitive lid opens automatically to accept waste, and then closes into a fitted well that helps to reduce odor and airborne contamination.
Use with SP Bel-Art super-strength or standard red Biohazard disposal bags, clear Biohazard disposal bags, Clavies® transparent autoclavable bags, or commercially available waste bags.