VWR® Disposal Carton for Plastics | Avantor
VWR® Disposal Carton for Plastics
63 L, 30.5L×30.5W×68.6H cm (12×12×27")
Catalog # 58949-700
Supplier:   VWR International
VWR® Disposal Carton for Plastics
Catalog # 58949-700
Supplier:  VWR International
VWR® Disposal Carton for Plastics
VWR® Disposal Carton for Plastics
63 L, 30.5L×30.5W×68.6H cm (12×12×27")
Price
$202.47
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 6
Product Details & Documents
Disposal boxes are shipped flat and fold into shape easily.
  • Clearly labeled, rigid cardboard carton; includes polypropylene liner to contain spills
  • Dedicated to separating plastics from the waste stream
  • Sized to fit under many benchtops
  • Approximate volume: 63 liters (16.6 gallons)
Entire box can be discarded when filled, ensuring safe handling and ready identification of contents.
Documents
Spécifications
  • Capacité:
    63 L
  • Dimensions:
    30.5L×30.5W×68.6H cm (12×12×27")