VITLAB® Dropping Bottles, LDPE, BrandTech
Fournisseur: Brandtech
Certificats
A propos de cet article
Translucent dropping bottles come with a high-density polyethylene dropper screw cap.
- Cut-to-size dropper with attached cap
- LDPE
- Five different sizes
The bottles provide excellent chemical resistance to aliphatic alcohols, aldehydes, alkalis, and acids.
Customers Who Bought This Also Bought