Amplitude™ Helix™ Wipes, Contec®
Fournisseur: CONTEC, INC. MX
Certificats
A propos de cet article
Amplitude™ Helix™ wipes are manufactured from meltblown polypropylene, combined with fine continuous filaments to provide improved cleanliness.
- Recommended for ISO Class 5 to 6 environments
- Continuous filaments provide low levels of particles and extractables
- Meltblown polypropylene allows metered release of disinfectants
- Microsized filaments provide exceptional particle removal
- Highly resistant to acids and chemicals
- Double packaged to preserve product integrity
Polypropylene has excellent acid and chemical resistance, making it ideal for use in many laboratory and cleanroom applications. The structure of the wipe is especially suitable for the metered release of solvents and disinfectants.
The microsized filaments provide exceptional particle removal. Manufactured solely from man-made fibers, the wipe has very low levels of particles and fibres as well as good sorbancy, and could be used for chemical spills.
Customers Who Bought This Also Bought