Amplitude™ Delta™ Wipes, Contec®
Fournisseur: Contec
Certificats
A propos de cet article
Amplitude™ Delta™ Wipes are made of 100% nonwoven polyester and manufactured using the spun lacing process with no binders or additives.
- 100% nonwoven polyester is ideal for ISO Class 5 to 6 environments
- Open fiber structure provides excellent sorptive capacity
- Durable nonwoven polyester is low in both particles and extractables
- 100% polyester is compatible with a wide range of solvents and disinfectants, including hypochlorites
- Double packaged to preserve product integrity
A cost effective polyester wipe with excellent cleanliness levels. The open structure makes it highly sorbent. It is a very soft wipe with both good wet and dry strength. 100% polyester construction offers good chemical resistance and compatibility with most solvents and disinfectants.
Customers Who Bought This Also Bought