Biohazard Waste Can Liner Bags, Justrite®
# de catalogue 56617-834
Fournisseur: Justrite
Spécifications
- Capacity:15 gal.
- Color:Red
- Description:Biohazard bags
- Thickness:0.4 mil
- Diameter ×H:838×610 mm
- Supplier No.:05901
Spécifications
A propos de cet article
For safe, temporary collection of biological waste or contaminated materials. Red color is easily identifiable as biohazard bags.
- 15 Gallon (56,8 l) capacity bag fits Justrite Biohazard cans of 6 and 10 gallon sizes
- High density PE of 11 microns thickness provides secure handling to avoid punctures and tears
- Tubular construction with a single side seal and star seal bottom provide a stable, leak proof bag
- Specifically labeled for Biohazard material handling
Printing includes English, Spanish and French translation along with the international recognizable biohazard symbol.
Customers Who Bought This Also Bought