KONTES® Fritted Hirsch Funnels, KIMBLE®, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Funnels feature a conical shape that directs filtrate through a reduced-area frit to concentrate crystal residue for rewashing.
Customers Who Bought This Also Bought