About this item
Medium-strength, removable, general-purpose Threadlocker 242™ adhesive is well suited for use with fasteners 6.4–19mm (¼–¾") in diameter.
- Medium-strength
- Removable
- Protects fasteners against vibration, loosening, leakage, and thread corrosion
Adhesive resists lubricants, solvents, and most chemicals. Parts can be separated using hand tools.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 19807-510
- 19807-514
- 19807-512
- Viscosity:
- 1200–5000 cP, thixotropic
- 1200–5000 cP, thixotropic
- 1200–5000 cP, thixotropic
- Torque:
- 12.4 N·m (110 in-lbs.) breakaway, 4.9 N·m (43 in-lbs.) prevail
- 12.4 N·m (110 in-lbs.) breakaway, 4.9 N·m (43 in-lbs.) prevail
- 12.4 N·m (110 in-lbs.) breakaway, 4.9 N·m (43 in-lbs.) prevail