VWR® Good Laboratory Practice Equipment Log Books, Pocket Size
Supplier: VWR International
Certificates
About this item
Pocket-size equipment log books feature user data, guidelines, and documentation pages.
- Designed to lay flat
- Number of pages: 96
- Cover color: red
Acid-free, archival-safe paper pages are sequentially numbered. Bound in a flexible imitation leather cover specially coated for durability in laboratory environments.
Customers Who Bought This Also Bought