BCR® Spiral Notebooks, Berkshire
Supplier: Berkshire
Certificates
About this item
Easy-to-use spiral notebooks feature either grid or college ruled formats
- Made with Berkshire cleanroom bond 1040 paper
- 100 pages/50 sheets per notebook
- Latex-free
- ISO Class 3–8
- Cleanroom Class 1–100,000
Notebooks have durable, solvent-resistant, translucent covers with 100 pages (50 sheets) printed on BCR® bond 1040 paper, bound by a noncontaminating polyethylene spiral.
Ordering information: Each notebook is individually wrapped. To customize notebooks with a company logo, contact your Avantor representative for more information.
Customers Who Bought This Also Bought