CoolSafe PCR Tube Cooling Chamber, Diversified Biotech
# de catalogue 89024-892
Fournisseur: Diversified Biotech
Small Business Enterprise
Spécifications
- Cat. no.:89024-892
- W×L×H:15.7 x 16.5 x 2.5 cm (63/16 x 61/2 x 1")
- Supplier No.:CHAM-1020
Spécifications
A propos de cet article
Anodized aluminum cooling chamber with 212 wells is ideal for temperature-sensitive liquid transfers
May be used when storing samples in ice buckets, freezers, and on hot plates. The chamber accommodates two 96-well PCR trays or 192 tubes sized 0.2mL, as well as twelve 0.5mL tubes and eight 1.5mL tubes.
Customers Who Bought This Also Bought