PYREX® Mallory Direct Reading Settlometer, Corning
# de catalogue 26307-604
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Capacity:2000 ml
- Description:Direct Reading Settlometer, Mallory
- Cat. no.:26307-604
- Supplier No.:7250-2L
Spécifications
A propos de cet article
Used with oxidized sludge process for treatment of sewage and other wastes. These cylinders are graduated in cc per litre and in hundredths of a foot.
Int.Ø×H: 130×190.5 mm
Customers Who Bought This Also Bought