Safety-Coated Boston Round Bottles, Amber, Qorpak
Fournisseur: Qorpak
Certificats
A propos de cet article
Safety-coated amber Boston round bottles protect samples from UV rays and are ideal for light sensitive products.
- Safety-coated boston round bottles are made of glass
If the glass breaks, the plastisol coating contains the substance in the bottle long enough to allow for proper disposal. These bottles are slip- and impact-resistant, making them the perfect bottle for handling volatile substances.
Ordering information: Bottles 89204-842, 93000-926, 93000-930, 93000-932, and 93000-954 are supplied Vacuum treated and Ionized. Bottle 89021-872 is supplied pre-cleaned for use with volatiles.
Customers Who Bought This Also Bought