Safety-Coated Packer Bottles, Wide Mouth, Amber, Qorpak
Fournisseur: Qorpak
Certificats
A propos de cet article
Reduce breakage and lab mishaps with these safety coated amber wide mouth packer bottles.
- Safety-coated packer bottles are made of glass
If the glass breaks, the plastisol coating contains the substance in the bottle long enough to allow for proper disposal.
Ordering information: Bottles 89096-730, 89096-732, 89096-734, 89096-736, 89096-738, and 89096-740 are supplied Ultra Cleaned. Bottles 93000-914, 93000-916, 93000-918, 93000-920, 89212-384, and 89212-386 are supplied vacuumed and ionized.
Customers Who Bought This Also Bought