A propos de cet article
PST® 565™ cures rapidly to provide low-pressure sealing for metal tapered pipe thread fittings.
- Cures rapidly
- Allows for ease of disassembly
- Withstands continuous temperatures to 149°C (300°F)
Sealant's controlled strength allows for ease of disassembly. Sealant is solvent-resistant, protects mated threaded areas from rust and corrosion, and withstands continuous temperatures to 149°C (300°F). It takes four hours to seal to operating pressure.