Bevco® Breva Economy Polyurethane Chairs
Fournisseur: BevCo Precision Manufacturing
Certificats
A propos de cet article
Durable polyurethane chairs at an affordable price.
- The black polyurethane seat and back are impervious to water, oil, and most chemicals
- Polyurethane is puncture-resistant and easy-to-clean
- Contoured back is fully adjustable encouraging proper spinal alignment
- Convenient handle makes moving the chair effortless
- Waterfall seat aids in lower body circulation, improving overall wellness
- Pneumatic height adjustment allows you to easily change your seat height
- Sturdy black nylon base
- 18" adjustable chrome footring standard on tall height models
- 300 lbs. weight capacity
- Standard environment
The Breva Series polyurethane chair is an excellent economic value option. The Breva will meet your most basic seating needs while exceeding your overall expectations. The Breva Series is the economical way to stay productive and comfortable. For instance, these durable chairs offer total body support with basic ergonomic features at Bevco's lowest price point. These chairs showcase a rugged, self-skinned, black polyurethane seat and back that resist stains and damage from punctures, grease, water, and most chemicals making it an excellent selection for industrial and institutional daily use.
Certifications: ANSI/BIFMA; GreenGuard Gold.
Ordering information: Five-year warranty.
Spécifications
- Catalog No:
- 76326-356
- 76326-358
- 76326-360
- 76326-362
- Seat/back material:
- Polyurethane
- Polyurethane
- Polyurethane
- Polyurethane
- Weight capacity:
- 300 lbs.
- 300 lbs.
- 300 lbs.
- 300 lbs.