ContactUsLinkComponent
Threadlocker 271™ High Strength Adhesive, Loctite®, Henkel
Threadlocker 271™ High Strength Adhesive, Loctite®, Henkel
# de catalogue 83007-504
Fournisseur:  Loctite
UL Listed
Threadlocker 271™ High Strength Adhesive, Loctite®, Henkel
# de catalogue 83007-504
Fournisseur:  Loctite
UL Listed
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    50 ml bottle
  • Supplier No.:
    27131
Spécifications
A propos de cet article
Designed for the permanent locking and sealing of fasteners such as large bolts and studs (M25 and larger) up to 25mm (1") diameter.
  • Acrylic threadlocker
  • High-strength
  • Low-viscosity
The adhesive cures when confined in the absence of air between close fitting metal surfaces and prevents loosening and leakage from shock and vibration. Low viscosity reduces the migration of liquid adhesive after application.
Certifications: Adhesive meets MIL-SPEC (S-46163A) for existing designs and ASTM D5363 for new designs. CFIA certified. UL listed.
Customers Who Bought This Also Bought