ContactUsLinkComponent
Super Bonder® 409™ Industrial Grade Instant Adhesive Gel, Loctite®, Henkel
Super Bonder® 409™ Industrial Grade Instant Adhesive Gel, Loctite®, Henkel
# de catalogue 83007-506
Fournisseur:  Loctite
Super Bonder® 409™ Industrial Grade Instant Adhesive Gel, Loctite®, Henkel
# de catalogue 83007-506
Fournisseur:  Loctite
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Super Bonder® 409™ industrial grade instant adhesive gel
  • Cat. no.:
    83007-506
  • Supplier No.:
    40945
Spécifications
A propos de cet article
Clear adhesive gel for general-purpose use on metal, plastic, and rubber.
  • Clear instant adhesive
  • Thixotropic gel formulation
  • Prevents adhesive runoff or drip
The thixotropic gel formulation fills large gaps and is useful for overhead or hard-to-reach applications where adhesive runoff or drip must be avoided.
Customers Who Bought This Also Bought