A propos de cet article
Lightweight, durable brush heads feature fusion-set fibers for maximum dependability and cleanliness
Perfex adjustable handle is made of high quality polyester and composite materials using pull-wound technology. Perfex adjustable handles are extremely durable , lightweight (16oz.) and designed to meet the hygienic needs of today's controlled environments. Smooth finish, impervious to water, grease, gasoline, detergents, sanitizers and solvents. Will not rot, corrode or oxidize. Widely accepted as a superior alternative to fixed length handles. Adjusts from 34” - 62”, compatible with gamma, ETO and autoclave sterilization. USDA and FDA Approved Materials.
Perfex adjustable handle is made of high quality polyester and composite materials using pull-wound technology. Perfex adjustable handles are extremely durable , lightweight (16oz.) and designed to meet the hygienic needs of today's controlled environments. Smooth finish, impervious to water, grease, gasoline, detergents, sanitizers and solvents. Will not rot, corrode or oxidize. Widely accepted as a superior alternative to fixed length handles. Adjusts from 34” - 62”, compatible with gamma, ETO and autoclave sterilization. USDA and FDA Approved Materials.
The stiff poly fiber texture is useful for cleaning rough unfinished surfaces. Lite-N-Tite® handle/broom head connection locks handle securely in place. Brush head will not warp, crack, or decay. Brush and handle are nonconductive. Will not absorb bacteria, liquids, or odors. Unaffected by water, petroleum products, detergents, sanitizers, and solvents.