Super Polx® 1200 Wipers, Berkshire
Supplier: Berkshire
Certificates
About this item
Durable, 100% knitted polyester wipers for general cleaning purposes, spill pickup, and equipment cleaning.
- Low particles, fibers, ions, and extractable
- High abrasion resistance
- Chemically compatible with IPA, acetone, and other solvents
- Soft hand and edge for sensitive surfaces
Continuous-filament, no-run construction offers resistance to chemicals and abrasion. Low extractables, with extremely low particle and fiber generation. For use in ISO Class 4 (FED-STD-209E Class 10/M2.5) environments. Manufactured and tested with strict adherence to quality assurance guidelines. Laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Specifications
- Catalog No:
- 21914-401
- 21914-474
- 21914-445
- Basis weight:
- 143 g/m²
- 143 g/m²
- 143 g/m²
- Absorbency - extrinsic:
- 426 ml/m²
- 426 ml/m²
- 426 ml/m²
- Absorbency - intrinsic:
- 3.7 ml/g
- 3.7 ml/g
- 3.7 ml/g
- Absorbency rate:
- 1 sec.
- 1 sec.
- 1 sec.
- Extractables:
- 0.016% using deionized water, 0.060% using isopropyl alcohol
- 0.016% using deionized water, 0.060% using isopropyl alcohol
- 0.016% using deionized water, 0.060% using isopropyl alcohol
- Fibres (>100 µm):
- 0.76/cm²
- 0.76/cm²
- 0.76/cm²
- Particle generation (>=0,5 µm) - biaxial shake:
- 1.4 x 10³/cm²
- 1.4 x 10³/cm²
- 1.4 x 10³/cm²
- Ions - calcium:
- 0.14 ppm
- 0.14 ppm
- 0.14 ppm
- Ions - magnesium:
- 0.031 ppm
- 0.031 ppm
- 0.031 ppm
- Ions - potassium:
- 0.0012 ppm
- 0.0012 ppm
- 0.0012 ppm
- Ions - sodium:
- 0.027 ppm
- 0.027 ppm
- 0.027 ppm