Polx® 1200 Wipers, Berkshire
Fournisseur: Berkshire
Certificats
A propos de cet article
Well-suited to optical applications and use on other sensitive surfaces, these all-purpose wipers are an economical midpoint between nonwoven materials and laundered synthetic wipers.
- Low particles, fibers, ions, and extractable
- High abrasion resistance
- Chemically compatible with IPA, acetone, and other solvents
- Soft hand and edge for sensitive surfaces
Well-suited to optical applications and use on other sensitive surfaces, these all-purpose wipers are an economical midpoint between nonwoven materials and laundered synthetic wipers.
Wipers are made of durable, yet nonabrasive 100% double-knit polyester. They are low in particle generation and extractables, making them useful in applications that would abrade or shred a nonwoven material. Recommended for ISO Class 5 (FED-STD-209E Class 100/3.5) environments.
Spécifications
- Catalog No:
- 21914-300
- 21914-354
- 10810-650
- 21914-332
- Basis weight:
- 101 g/m²
- 101 g/m²
- 101 g/m²
- 101 g/m²
- Absorbency - extrinsic:
- 363 ml/m²
- 363 ml/m²
- 363 ml/m²
- 363 ml/m²
- Absorbency - intrinsic:
- 3.6 ml/g
- 3.6 ml/g
- 3.6 ml/g
- 3.6 ml/g
- Absorbency rate:
- 1 sec.
- 1 sec.
- 1 sec.
- 1 sec.
- Extractables:
- 0.025% using deionized water, 0.090% using isopropyl alcohol
- 0.025% using deionized water, 0.090% using isopropyl alcohol
- 0.025% using deionized water, 0.090% using isopropyl alcohol
- 0.025% using deionized water, 0.090% using isopropyl alcohol
- Fibres (>100 µm):
- 10/cm²
- 10/cm²
- 10/cm²
- 10/cm²
- Particle generation (>=0,5 µm) - biaxial shake:
- 13 x 10³/m²
- 13 x 10³/m²
- 13 x 10³/m²
- 13 x 10³/m²