Quality Control in Analytical Chemistry, 2nd Edition, JAY C. WILEY
# de catalogue 56615-568
Fournisseur: JAY C. WILEY
Spécifications
- Description:Quality Control in Analytical Chemistry, 2nd Edition by G. Kateman
- Cat. no.:56615-568
- Supplier No.:0471-55777-3
Spécifications
A propos de cet article
This edition includes numerous modern methods and the latest developments in time-proven techniques. Written for physical chemists and chemical engineers.
- Describes the basics of analytical techniques, sampling, and data handling in order to improve quality control in analytical laboratory management
- Stresses which quality parameters can be improved and which ones should be rectified first
336 pages, released 1993
Customers Who Bought This Also Bought