ValuSeal® 1500 Sealed-Edge Cleanroom Wipers, Berkshire
Fournisseur: Berkshire
Certificats
A propos de cet article
These 100% knitted polyester wipers feature double-knit construction, which makes them highly absorbent, capable of holding more liquid than many polyester knits.
- Low particles, fibers, ions, and extractable
- Chemically compatible with IPA, acetone, and other solvents
- Lightweight, strong, abrasion- and tear-resistant
- High absorbency material reduces actual cost in use compared to heavier weight products
Sealed edges ensure extremely low levels of particles, fibers, ions, and residues. For use in ISO Class 3 (FED-STD-209E Class 1/M1.5) environments.
Wipers are laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom, minimizing the potential for contamination.
Spécifications
- Catalog No:
- 47730-818
- 47727-366
- Basis weight:
- 125 g/m²
- 125 g/m²
- Absorbency - extrinsic:
- 498 ml/m²
- 498 ml/m²
- Absorbency - intrinsic:
- 4 ml/g
- 4 ml/g
- Absorbency rate:
- 1 sec.
- 1 sec.
- Extractables:
- 0.015% using deionized water, 0.077% using isopropyl alcohol
- 0.015% using deionized water, 0.077% using isopropyl alcohol
- Fibres (>100 µm):
- 0.033/cm²
- 0.033/cm²
- Particle generation (>=0,5 µm):
- 1.7 x 10³/cm²
- 1.7 x 10³/cm²