VWR® Micro Spoon, Stainless Steel
# de catalogue 82027-526
Fournisseur: VWR International
Spécifications
- Length:16.5 cm (61/2")
- Cat. no.:82027-526
Spécifications
A propos de cet article
Designed for use in the transfer and handling of small chemical or biological samples.
- Rustproof
- Stainless steel construction
The rustproof micro spoon is constructed of stainless steel and features a plastic handle. Substances will not stick to spoon surface.
Customers Who Bought This Also Bought