SP Bel-Art Economy Nonsterile Sample Spoons, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
These economical measuring devices are ideal for retrieving small, specific volume samples of powders or liquids.
- Economical
- Unique Flat-bottom shape
- Nonsterile
- White FDA-grade polystyrene construction
Their unique flat-bottom shape allows them to be used as weighing boats for additional sampling accuracy.
Use a separate spoon for each material sampled to avoid cross contamination. All sizes are made from white, FDA-grade high-impact polystyrene plastic for durability and stiffness when sampling packed powders.
Ordering information: Supplied 12 per bag.
Customers Who Bought This Also Bought