VWR® Spec-Wipe® 7 Wipers
Fournisseur: VWR International
Certificats
A propos de cet article
Excellent for cleaning rough, abrasive or irregular surfaces, yet soft enough for delicate surfaces.
- Sealed edges
- No-run interlock knit provides durability for vigorous cleaning
- Edges are sealed for reduced lint and fiber
Excellent for cleaning rough, abrasive or irregular surfaces, yet soft enough for delicate surfaces.
Use these 100% interlock knit polyester wipers where low particle and fiber contamination are most critical.
Appropriate for use in electronic, semiconductor, aerospace, pharmaceutical, medical device, research, and education industries. Laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom to further reduce particulate and extractable contaminants.
Certifications: Laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Spécifications
- Catalog No:
- 21913-216
- 21913-214
- Basis weight:
- 136 g/m²
- 136 g/m²
- Absorbency:
- 312 ml/m²
- 312 ml/m²
- Absorbency rate:
- <1.5 sec.
- <1.5 sec.
- Extractables:
- 0.015 g/m² using deionized water, 0.015 g/m² using isopropyl alcohol
- 0.015 g/m² using deionized water, 0.015 g/m² using isopropyl alcohol
- Fibres (>100 µm):
- 0.6×10³/m²
- 0.6×10³/m²
- Particle generation (>=0,5 µm):
- 4.8 x 10⁶/m²
- 4.8 x 10⁶/m²
- Ions - chloride:
- 0.13 ppm
- 0.13 ppm
- Ions - sodium:
- 0.11 ppm
- 0.11 ppm