SP Bel-Art Tech Pens, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
These heavy-duty, soft-tip disposable pens write on glass, ceramics, metal, plastics, and paper.
- Heavy-duty, disposable design
- Snap-on caps indicate ink color
- Autoclavable
- Soft tip writes on glass, ceramics, metal, plastics, and paper
The oil-based marking paint resists washing, weathering, mild acids, and alkalies. Pens withstand autoclaving at 121°C (250°F). Pens operate within the temperature range of -46 to 65°C (-50 to 150°F). Mark is approximately 1 mm (0.040") wide.
Frequently Bought Together