Solvent-Based Paint Pen Markers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Writes on all plastics, glass, metal, wood, paper and cloth, including Teflon® fluoropolymer resins.
- Precision seal valve provides smooth ink flow and preserves the pen’s life, even if left uncapped
- When dry, the solvent based ink is temperature resistant from 0 to 155 °C (32 to 310 °F)
- Markings are insoluble in water or hydrocarbons
Resists many solvents, but is easily removed with alcohol or acetone.
Customers Who Bought This Also Bought