ValuSeal-HA® Sealed-Edge Cleanroom Wipers, Berkshire
Fournisseur: Berkshire
New Product
Certificats
A propos de cet article
Highly absorbent, 100% knitted polyester wipers have ultrasonic heat-sealed edges.
- Lightweight
- Low particles, fibers, ions, and extractable
- High absorbency material reduces actual cost in use compared to heavier weight products
Wipers work well with IPA, acetone, and other solvents. Steam autoclavable for aseptic environments.
Certifications: Recommended for ISO Class 3 (FED-STD-209E Class 1/M1.5) cleanroom environments, including semiconductor and biomedical cleanroom applications.
Ordering information: Wipers are laundered and packed in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Spécifications
- Catalog No:
- 80094-522
- 80094-518
- Basis weight:
- 129 g/m²
- 129 g/m²
- Absorbency - extrinsic:
- 474 ml/m²
- 474 ml/m²
- Absorbency - intrinsic:
- 3.7 ml/g
- 3.7 ml/g
- Absorbency rate:
- 1 sec.
- 1 sec.
- Extractables:
- 0.012% using deionized water, 0.05% using isopropyl alcohol
- 0.012% using deionized water, 0.05% using isopropyl alcohol
- Fibres (>100 µm):
- 0.045/cm²
- 0.045/cm²
- Particle generation (>=0,5 µm) - biaxial shake:
- 0.7×10³/cm²
- 0.7×10³/cm²