VWR® Annealing Cup, Porcelain
# de catalogue 89038-212
Fournisseur: VWR International
Spécifications
- Capacity:15 ml
- Cat. no.:89038-212
- O.D.×H:38 x 25 mm
Spécifications
A propos de cet article
Chemical-porcelain annealing cup is glazed inside and out, except for outside bottom surface and rim.
- Conical form
- Glazed interior and exterior except for bottom and rim
- Chemical-porcelain construction
Smooth interior surfaces in a conical form.
Customers Who Bought This Also Bought