VWR® Bitumen Crucible, Porcelain
# de catalogue 89038-036
Fournisseur: VWR International
Spécifications
- Capacity:28 ml
- Cat. no.:89038-036
- O.D.×H:44 x 24 mm (13/4 x 15/16")
Spécifications
A propos de cet article
Chemical-porcelain bitumen crucible features a perforated bottom and large filtering surface.
- Glazed inside and out
- Used with 30 to 33 mm diameter filter paper
Crucible is glazed inside and out, except for outside bottom surface and rim. For use with 30–33mm diameter filter paper.
Customers Who Bought This Also Bought