VWR® Crucibles, PTFE
Fournisseur: VWR International
Certificats
A propos de cet article
Isostatically molded, pure FDA approved PTFE crucibles are totally chemically resistant and inert.
- Chemically resistant
- Inert
- PTFE fabrication
For use in temperatures up to 270°C (518°F).
Customers Who Bought This Also Bought