Walter Crucible Holder, PLASTICOID COMPANY INC
# de catalogue 24065-000
Fournisseur: Plasticoid
Small Business Enterprise
Spécifications
- Description:Walter Crucible Holder
- Cat. no.:24065-000
- Supplier No.:VCHWGS
Spécifications
A propos de cet article
Rubber crucible holder for use with Gooch crucibles, or similar crucibles, in the neck of a standard suction flask.
- For use with crucibles 20 to 35 mL, and flasks 250 to 1000 mL
Delivery: Supplied with funnel.
Customers Who Bought This Also Bought