Clay Annealing Cups, DFC Ceramics
Fournisseur: DFC Ceramics
Certificats
A propos de cet article
Cups are used for annealing gold buttons and for ignition of precipitates, such as barium sulfate, copper sulfide, silica, etc.
- Smooth, unglazed surface
- Supplied without cover
Customers Who Bought This Also Bought