Packer Bottles, Amber, Wide Mouth
Fournisseur: Qorpak
Certificats
A propos de cet article
Amber wide mouth Packer bottles feature a wide opening making them the ideal general-purpose bottle for solids.
- Packer bottles are made of glass
- Ideal for light sensitive products
Applications include pharmaceutical, chemical, and environmental.
Ordering information: Bottles 11311-528, 11311-530, 11311-532, 11311-534, 11311-540, 10158-494 and 10158-496 are supplied Ultra Cleaned. Bottles 36319-156, 36319-158, 36319-250, 36319-253, 36319-256, 36319-265, 36319-268, and are supplied pre-cleaned for volatiles. Bottles 89204-848, BA11311-362CRC, 76487-192, 76487-194, 76487-196, 76487-198 and 76487-200 are supplied pre-cleaned and certified for semi-volatiles.
Customers Who Bought This Also Bought