Standard Wide Mouth Bottles, Clear, Qorpak
Fournisseur: Qorpak
Certificats
A propos de cet article
Clear, flint glass standard wide mouth bottles feature a wide opening making them the ideal general-purpose bottle.
Applications include pharmaceutical, chemical, environmental, food packaging and food storage.
Ordering information: Bottles 36319-155, 36319-195, 36319-199, 36319-219, 36319-225, and 89204-692 are supplied pre-cleaned for volatiles. Bottles 89176-848, 89176-850, 93001-560, 93001-562, 16054-206, 76487-214 and 10053-746 are supplied pre-cleaned and certified for semi-volatiles.
Customers Who Bought This Also Bought